missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNLZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566527
This item is not returnable.
View return policy
Description
Immunogen sequence: LGWFSDLNGKR NIEEILTARG EQVHRVAGEG ALELVLEAAG APTSTAAPEA GE Highest antigen sequence identity to the following orthologs - mouse 64%, rat 70%.
May function as a co-chaperone towards HSPA9/mortalin which, by itself, is prone to self-aggregation.
Specifications
| DNLZ | |
| Polyclonal | |
| Unconjugated | |
| DNLZ | |
| bA413M3.2; C9orf151; DNL-type zinc finger; DNL-type zinc finger protein; DNLZ; HEP; HEP1; HSP70 escort protein; hsp70-escort protein 1; mtHsp70-escort protein; RP11-413M3.2; TIMM15; translocase of inner mitochondrial membrane 15 homolog; ZIM17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 728489 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5SXM8 | |
| DNLZ | |
| Recombinant Human DNLZ. Recombinant protein control fragment (Product #RP-105240). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction