missing translation for 'onlineSavingsMsg'
Learn More

DNAL1 Antibody (29E4), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18797471 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18797471 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18797471 Supplier Novus Biologicals Supplier No. NBP242657MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNAL1 Monoclonal antibody specifically detects DNAL1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAL1
Applications Flow Cytometry
Classification Monoclonal
Clone 290000
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias C14orf168MGC12435,1700010H15RiK, chromosome 14 open reading frame 168, dynein light chain 1, axonemal, dynein, axonemal, light chain 1
Host Species Mouse
Immunogen Recombinant human DNAL1 (1-190aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83544
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.