Learn More
DNAJC14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553618
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: WGWLELPWVK QNINRQGNAP VASGRYCQPE EEVARLLTMA GVPEDELNPF HVLGVEATAS DVELKKAYRQ LAVMVHPDKN HHPRAEEAFK VLRAAWDIVS NAEKRKEYEM KRMAENELSR SVNEFLSKLQ DDLKEAMNTM MCSRCQG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
ER-membrane-associated protein; may be involved in ER export regulation of a GPCR.Specifications
DNAJC14 | |
Polyclonal | |
Unconjugated | |
DNAJC14 | |
5730551F12Rik; DNAJ; DnaJ (Hsp40) homolog, subfamily C, member 14; DnaJ heat shock protein family (Hsp40) member C14; dnaJ homolog subfamily C member 14; DnaJ protein homolog 3; Dnajc14; dopamine receptor interacting protein; dopamine receptor-interacting protein of 78 kDa; DRiP78; HDJ3; hDj-3; Human DnaJ protein 3; LIP6; LYST-interacting protein LIP6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114481, 74330, 85406 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5XIX0, Q6Y2X3, Q921R4 | |
DNAJC14 | |
Recombinant protein corresponding to Human DNAJC14 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only