missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | DNAJC10 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16841154
|
Proteintech
13101-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16831154
|
Proteintech
13101-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ERdj5 antibody displays a higher affinity for endogenous ERdj5u.
This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.Specifications
| DNAJC10 | |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q498R3, Q8IXB1, Q9DC23 | |
| 295690, 54431, 66861 | |
| DNAJC10 Fusion Protein Ag3737 | |
| Primary | |
| -20°C | |
| DNAJC10 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DNAJC10, ER resident protein ERdj5, ERdj5, JPDI, Macrothioredoxin, MTHr | |
| DNAJC10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title