missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: VLGLQRYASP EDIKKAYHKV ALKWHPDKNP ENKEEAERKF KEVAEAYEVL SNDEKRDIYD KYGTEGLNGG GSHFDDECEY GFTFHKPDDV FKEIFHERDP FSFHFFEDSL EDLLNRPGSS YGNRNRDAGY FFSTASEYP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.
DNAJB7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).
Specifications
Specifications
| Antigen | DNAJB7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Dnajb7 |
| Gene Accession No. | Q7Z6W7 |
| Gene Alias | 4933424H20Rik; DJ5; DnaJ (Hsp40) homolog, subfamily B, member 7; DnaJ heat shock protein family (Hsp40) member B7; dnaJ homolog subfamily B member 7; Dnajb7; dnaJ-like 5; HSC3; mDj5; novel DnaJ domain protein |
| Gene Symbols | Dnajb7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
