Learn More
DNAJB2 Monoclonal Antibody (OTI5E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502263
Description
The DnaJ family is one of the largest of all the chaperone families and has evolved with diverse cellular localization and functions. The presence of the J domain defines a protein as a member of the DnaJ family. DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are involved in the regulation of protein degradation, exocytosis and endocytosis. DnaJB2 (DnaJ homolog subfamily B member 2), also known as HSJ1 or HSPF3, is expressed almost exclusively in the brain, with the highest levels in the frontal cortex and hippocampus. Two isoforms are produced due to alternative splicing.Specifications
| DNAJB2 | |
| Monoclonal | |
| 0.48 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P25686 | |
| DNAJB2 | |
| Full length human recombit protein of human DNAJB2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI5E5 | |
| Unconjugated | |
| DNAJB2 | |
| CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3300 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.