Learn More
DNAH12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557974
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IHGLYLDGAR WDRESGLLAE QYPKLLFDLM PIIWIKPTQK SRIIKSDAYV CP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
DNAH12 gene ontology annotations related to this gene include microtubule-based movement.Specifications
DNAH12 | |
Polyclonal | |
Unconjugated | |
DNAH12 | |
4921531P07Rik; axonemal beta dynein heavy chain 12; Axonemal dynein heavy chain 12-like protein; Axonemal dynein heavy chain 7-like protein; axonemal dynein heavy chain isotype3; Bm259; ciliary dynein heavy chain 12; DHC3; DLP12; Dnah12; DNAH12L; DNAH7L; DNAHC12; Dnahc3; Dnahc7c; Dnahc7l; DNHD2; dynein axonemal heavy chain 12; dynein heavy chain 12, axonemal; dynein heavy chain 12, axonemal-like; Dynein heavy chain 7-like, axonemal; dynein heavy chain domain 2; dynein heavy chain domain-containing protein 2; dynein, axonemal, heavy chain 12; dynein, axonemal, heavy polypeptide 12; dynein, heavy chain-5; Gm284; Gm74; Gm907; Hdhc3; HL19; HL-19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201625 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZR08 | |
DNAH12 | |
Recombinant protein corresponding to Human DNAH12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only