Learn More
STEMCELL Technologies DNA Loading Dye
Product Code. 17179901
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 1 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17179901 3 x 1 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17179901 Supplier STEMCELL Technologies Supplier No. 79018
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Ready-to-use marker dye for loading DNA samples into agarose gel wells and tracking migration during electrophoresis
  • 1 kb DNA Ladder allows for determining the size of double-stranded DNA from 250 - 10000 base pairs (bp) by gel electrophoresis
  • The ladder consists of 13 double-stranded, blunt-end DNA fragments, of sizes 250, 500, 750, 1000, 1500, 2000, 2500, 3000, 4000, 5000, 6000, 8000, and 10,000 bp (see Figure 1)
  • The 1000 and 3000 bp fragments have greater intensity relative to the other bands for easy identification
  • DNA Ladder is not intended for quantitative analysis
  • The 1 kb DNA ladder comes pre-mixed with DNA Loading Dye, which is also available for individual sale (Catalog #79018), for loading DNA samples into agarose gel wells
  • The recommended loading volume of the 1 kb DNA ladder is 6 μL per lane
Specifications
For Use With (Application) Disease Modeling
Quantity 3 x 1 mL