Learn More
DMRTC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21473-1-AP-20UL
 View more versions of this product
Product Code. 16871065
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
DMRTC1B, also named as Doublesex- and mab-3-related transcription factor C1, is a 192 amino acid protein, which belongs to the DMRT family. DMRTC1B is predomitly expressed in kidney, pancreas, ovary and testis. Detected in brain and in many other tissues. DMRTC1B exists two isoform and molecular weight of isoforms are 20 kDa and 13 kDa.
Specifications
Specifications
DMRTC1B
Polyclonal
Unconjugated
DMRTC1B
DMRT like family C1B, DMRTC1, DMRTC1B
Rabbit
Antigen Affinity Chromatography
RUO
632708, 728656
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5HYR2
Dmrtc1b
DMRTC1B Fusion Protein Ag15419
20 μL
Primary
Human, Mouse