missing translation for 'onlineSavingsMsg'
Learn More

DMPO Nitrone Adduct Antibody (N1664A), FITC, Novus Biologicals™

Product Code. 18232495 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18232495 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18232495 Supplier Novus Biologicals Supplier No. NBP259306F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DMPO Nitrone Adduct Monoclonal specifically detects DMPO Nitrone Adduct in Mouse, Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMPO Nitrone Adduct
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone N1664A
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias 2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide, 5,5-Dimethyl-1-pyrroline N-oxide, 5,5-Dimethyl-1-pyrroline-1-oxide
Host Species Mouse
Immunogen 5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA.
Target Species Mouse, Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.