missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMP4 Polyclonal Antibody

Description
DMP4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | DMP4 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | FAM20C |
| Gene Accession No. | Q8IXL6 |
| Gene Alias | C76981; Dentin matrix protein 4; DMP4; DMP-4; Extracellular serine/threonine protein kinase FAM20C; Fam20c; family with sequence similarity 20 member C; family with sequence similarity 20, member C; GEF-CK; Golgi casein kinase; golgi-enriched fraction casein kinase; Kiaa4081; mKIAA4081; RGD1311980; RNS |
| Gene Symbols | FAM20C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?