missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMBX1 Monoclonal Antibody (1A9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524393
This item is not returnable.
View return policy
Description
Peptide Sequence: MQHYGVNGYS LHAMNSLSAM YNLHQQAAQQ AQHAPDYRPS VHALTLAERL AGCTFQDIIL EARYGSQHRK QRRSRTAFTA QQLEALEK.
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| DMBX1 | |
| Monoclonal | |
| Unconjugated | |
| DMBX1 | |
| Atx; Cdmx; diencephalon/mesencephalon homeobox 1; diencephalon/mesencephalon homeobox protein 1; Diencephalon/mesencephalon-expressed brain homeobox gene 1 protein; dmbx1; homeobox gene Atx; homeoprotein MBX; MBX; Orthodenticle homolog 3; OTX; OTX3; Paired-like homeobox protein DMBX1; paired-type homeobox Atx; PaxB | |
| Mouse | |
| Protein A | |
| RUO | |
| 127343 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 1A9 | |
| PBS with no preservative; pH 7.4 | |
| Q8NFW5 | |
| DMBX1 | |
| DMBX1 (NP_671725, 1 a.a. ~ 88 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction