missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143408
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 2.
Specifications
| DLX2 | |
| Polyclonal | |
| Unconjugated | |
| AW121999; DII A; distal-less homeo box 2; distal-less homeobox 2; DLX2; Dlx-2; Dlx5; DLX-5; homeobox Dlx5; homeobox protein DLX-2; Homeobox protein TES-1; TES1; Tes-1 | |
| Recombinant protein encompassing a sequence within the center region of zebrafish Dlx2a. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction