missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572199
This item is not returnable.
View return policy
Description
DLK1 Polyclonal Antibody for Western Blot, IHC (P), Flow
The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
Specifications
| DLK1 | |
| Polyclonal | |
| Unconjugated | |
| DLK1 | |
| Adipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOG | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 13386, 8788 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.05% ProClin 300; pH 7.4 | |
| P80370, Q09163 | |
| DLK1 | |
| KLH conjugated synthetic peptide between 368-402 amino acids from the C-terminal region of human DLK1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction