Learn More
DLG5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564881
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: FLHKPFPGGPL QVCPQACPSA SERSLSSFRS DASGDRGFGL VDVRGRRPLL PFETEVGPCG VGEASLDKAD SEGSNSGGTW PKAMLSSTAV PEKLSVYKKP KQRKSI Highest antigen sequence identity to the following orthologs - mouse 76%, rat 73%.
May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.Specifications
DLG5 | |
Polyclonal | |
Unconjugated | |
Dlg5 | |
4933429D20Rik; discs large homolog 5; discs large MAGUK scaffold protein 5; discs large protein LP-DLG; discs large protein P-dlg; discs, large homolog 5; discs, large homolog 5 (Drosophila); disks large homolog 5; DLG5; KIAA0583; large type of P-DLG; LP-DLG; mKIAA0583; PDLG; P-DLG5; Placenta and prostate DLG; T25557 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDM6 | |
Dlg5 | |
Recombinant Human DLG5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only