missing translation for 'onlineSavingsMsg'
Learn More

DLG1 Antibody (S06-6C8), Novus Biologicals™

Product Code. 18690995 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18690995 50 μg 50µL
18624384 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18690995 Supplier Novus Biologicals Supplier No. NBP32000250ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DLG1 Monoclonal antibody specifically detects DLG1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLG1
Applications Western Blot
Classification Monoclonal
Clone S06-6C8
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97
Host Species Rabbit
Immunogen A synthetic peptide of human DLG1 (Uniprot # Q12959)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1739
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.