missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLEC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538830
Additional Details : Weight : 0.00900kg
Description
This gene contains 37 exons, spans approximately 59-kb, and is located in the 3p22-p21.3 chromosomal segment that is commonly deleted in various carcinomas. Several alternatively spliced transcripts have been observed that contain disrupted coding regions and likely encode nonfunctional proteins. Aberrant transcription of this gene may be involved in carcinogenesis of the lung, esophagus, and kidney. [provided by RefSeq, Jul 2008].Specifications
DLEC1 | |
Polyclonal | |
Unconjugated | |
DLEC1 | |
DLC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9940 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y238 | |
DLEC1 | |
A synthetic peptide derived from the N-terminal region of human DLEC1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |