missing translation for 'onlineSavingsMsg'
Learn More

DL-threo-2-methylisocitrate sodium, MedChemExpress

Product Code. 16491619
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491619 1 mg 1mg
16489627 5 mg 5mg
16437088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491619 Supplier Medchem Express Supplier No. HY16581A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DL-threo-2-methylisocitrate is a substrate of isocitrate lyase 1(ICL1).

DL-threo-2-methylisocitrate is a substrate of isocitrate lyase 1(ICL1).

Specifications

Chemical Name or Material DL-threo-2-methylisocitrate sodium
Physical Form Solid
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C14H14Na6O14
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 100 mg/mL (183.76 mM; Need ultrasonic) ∣
SMILES O[C@@](C)(C([O-])=O)[C@@]([H])(C([O-])=O)CC([O-])=O.O[C@](C)(C([O-])=O)[C@]([H])(C([O-])=O)CC([O-])=O.[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
Molecular Weight (g/mol) 544.19
Formula Weight 544.19
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.