Learn More
DL-Phenylalanine-3-13C, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Phenylalanine-13C (propanoic-3-13C acid) |
| CAS | 286425-42-1 |
| Molecular Formula | C8(13C)H11NO2 |
| Quantity | 25 mg |
| Synonym | Phenylalanine-β-13C (9CI), DL-Phenylalanine-β-13C, DL-Phenylalanine-13C, DL-Phenylalanine-13C (propanoic-3-13C acid), DL-[13C]Phenylalanine, 2-Amino-3-phenylpropanoic-3-13C acid, (RS)-Phenylalanine-13C |
| InChI Formula | InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/i6+1 |
| SMILES | NC(C(O)=O)[13CH2]C1=CC=CC=C1 |
| IUPAC Name | 2-amino-3-phenyl(3^{13}C)propanoic acid |
| Molecular Weight (g/mol) | 166.18 |
| Formula Weight | 166.08 |