Learn More
DL-Phenyl-d5-alanine, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Phenylalanine-D5 (phenyl-D5) |
| CAS | 284664-89-7 |
| Molecular Formula | C9H6D5NO2 |
| Quantity | 50 mg |
| Synonym | Phenyl-d5-alanine (9CI) |
| InChI Formula | InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/i1D,2D,3D,4D,5D |
| SMILES | O=C(C(CC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])N)O |
| IUPAC Name | 2-Amino-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 170.22 |
| Formula Weight | 170.11 |