Learn More
DL-Penicillamine, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Penicillamine |
| CAS | 52-66-4 |
| Molecular Formula | C5H11NO2S |
| Quantity | 1 g |
| Synonym | 3-Mercapto-DL-valine, (±)-Penicillamine, 2-Amino-3-mercapto-3-methylbutyric Acid, 3,3-Dimethylcysteine, DL-3-Mercaptovaline, DL-beta-Mercaptovaline, DMC, NSC 22880, NSC 44656, dl-DMC, beta,beta-Dimethylcysteine, beta-Mercaptovaline, beta-Thiovaline |
| InChI Formula | InChI=1S/C5H11NO2S/c1-5(2,9)3(6)4(7)8/h3,9H,6H2,1-2H3,(H,7,8) |
| SMILES | CC(C)(S)C(N)C(=O)O |
| IUPAC Name | 2-amino-3-methyl-3-sulfanylbutanoic acid |
| Molecular Weight (g/mol) | 149.21 |
| Formula Weight | 149.051 |