Learn More
DL-Penicillamine, TRC
Product Code. 30416752
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416752 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416752 Supplier Toronto Research Chemicals Supplier No. TRCP22301025G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Penicillamine
CAS 52-66-4
Molecular Formula C5H11NO2S
Quantity 25 g
Synonym 3-Mercapto-DL-valine, (±)-Penicillamine, 2-Amino-3-mercapto-3-methylbutyric Acid, 3,3-Dimethylcysteine, DL-3-Mercaptovaline, DL-beta-Mercaptovaline, DMC, NSC 22880, NSC 44656, dl-DMC, beta,beta-Dimethylcysteine, beta-Mercaptovaline, beta-Thiovaline
InChI Formula InChI=1S/C5H11NO2S/c1-5(2,9)3(6)4(7)8/h3,9H,6H2,1-2H3,(H,7,8)
SMILES CC(C)(S)C(N)C(=O)O
IUPAC Name 2-amino-3-methyl-3-sulfanylbutanoic acid
Molecular Weight (g/mol) 149.21
Formula Weight 149.051