Learn More
DL-Menthol, TRC
Product Code. 30011308
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011308 250 mg 250mg
17904875 1 g 1g
17921535 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011308 Supplier Toronto Research Chemicals Supplier No. TRCM218900250MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-78-1
Molecular Formula C10 H20 O
Molecular Weight (g/mol) 156.27
Synonym Menthol racemic, (1RS,2SR,5RS)-5-methyl-2-(1-methylethyl)cyclohexanol, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, (1R,2S,5R)-rel-, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, (1α,2β,5α)-, Menthol, cis-1,3,trans-1,4- (8CI), Menthol, dl- (6CI), (1R,2S,5R)-rel-5-Methyl-2-(1-methylethyl)cyclohexanol, (±)-Menthol, DL-Menthol, Fisherman's Friend Lozenges, Hexahydrothymol, Meggezone, Menthacamphor, Menthol, Menthomenthol, NSC 2603, Peppermint camphor, Racementhol, Racemic menthol, Therapeutic Mineral Ice, Thymomenthol, dl-Menthol, rac-Menthol, rel-(1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1
Formula Weight 156.1514
Chemical Name or Material Menthol Racemic