Learn More
DL-Lanthionine, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Lanthionine |
| CAS | 3183-08-2 |
| Molecular Formula | C6 H12 N2 O4 S |
| Quantity | 10 mg |
| Synonym | D-Cysteine, S-[(2S)-2-amino-2-carboxyethyl]-, rel-, Alanine, 3,3'-thiodi-, DL- (8CI), Cysteine, S-(2-amino-2-carboxyethyl)- (9CI), DL-Cysteine, S-(2-amino-2-carboxyethyl)-, rel-S-[(2S)-2-Amino-2-carboxyethyl]-D-cysteine, DL-Lanthionine |
| InChI Formula | InChI=1S/C6H12N2O4S/c7-3(5(9)10)1-13-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1 |
| SMILES | N[C@@H](CSC[C@H](N)C(=O)O)C(=O)O |
| IUPAC Name | (2R)-2-amino-3-[(2R)-2-amino-2-carboxyethyl]sulfanylpropanoic acid |
| Molecular Weight (g/mol) | 208.24 |
| Formula Weight | 208.0518 |