Learn More
DL-Cystine, TRC
Product Code. 17937824
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937824 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17937824 Supplier Toronto Research Chemicals Supplier No. TRCC9978501G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 923-32-0
Molecular Formula C6H12N2O4S2
Synonym NSC 203781
InChI Formula InChI=1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)
SMILES NC(CSSCC(N)C(=O)O)C(=O)O
IUPAC Name 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
Molecular Weight (g/mol) 240.3
Quantity 1 g
Formula Weight 240.0238
Shipping Condition Room Temperature
Chemical Name or Material DL-Cystine