missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. p-7038203
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443658 250 mg 250mg
15433658 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443658 Supplier Thermo Scientific Alfa Aesar Supplier No. H57113.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80028-44-0
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD14585012
InChI Key AYYKCALHTPKNOI-UHFFFAOYSA-N
Synonym ethyl pyrrolidine-3-carboxylate hydrochloride, dl-beta-proline ethyl ester hydrochloride, ethyl pyrrolidine-3-carboxylate hcl, 3-pyrrolidinecarboxylic acid, ethyl ester, hydrochloride, pubchem18711, ethylpyrrolidine-3-carboxylate hydrochloride, +/--pyrrolidine-3-carboxylic acid ethyl ester hy, 3-pyrrolidinecarboxylic acid ethyl ester hydrochloride
PubChem CID 12896003
IUPAC Name ethyl pyrrolidine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCNC1.Cl

Specifications

Quantity 250 mg
Formula Weight 179.65
Percent Purity 97%
Chemical Name or Material DL-beta-Proline ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.