missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-4-phosphonobutyric acid, 95%

Product Code. p-7047125
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11378616

Brand: Thermo Scientific Alfa Aesar L17671.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20263-07-4 | C4H10NO5P | 183.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20263-07-4
Molecular Formula C4H10NO5P
Molecular Weight (g/mol) 183.10
MDL Number MFCD00013999
InChI Key DDOQBQRIEWHWBT-UHFFFAOYNA-N
Synonym 2-amino-4-phosphonobutyric acid, dl-ap4, dl-2-amino-4-phosphonobutyric acid, butanoic acid, 2-amino-4-phosphono, +/--2-amino-4-phosphonobutyric acid, butyric acid, 2-amino-4-phosphono, 2-amino-4-phosphono-butyric acid, l-2-amino-4-phosphonobutyrate, l-apb, abu p
PubChem CID 2207
IUPAC Name 2-amino-4-phosphonobutanoic acid
SMILES NC(CCP(O)(O)=O)C(O)=O

Specifications

Melting Point 225°C to 228°C
Quantity 250 mg
Beilstein 2093908
Formula Weight 183.1
Percent Purity 95%
Chemical Name or Material DL-2-Amino-4-phosphonobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.