missing translation for 'onlineSavingsMsg'
Learn More

DKM 2-93, MedChemExpress

Product Code. 16400946
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16400946

Brand: Medchem Express HY101836

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress DKM 2-93 is a relatively selective inhibitor of UBA5 with an IC50 of 430 μM.

DKM 2-93 is a relatively selective inhibitor of UBA5 with an IC50 of 430 μM.

Specifications

Chemical Name or Material DKM 2-93
CAS 65836-72-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14ClNO3
Quantity 200 mg
Solubility Information DMSO : 125 mg/mL (512.95 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=C(OC)C(OC)=C1)CCl
Molecular Weight (g/mol) 243.69
Formula Weight 243.69
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.