missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DKK3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA580553

Product Code. 16377664

  • 341.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DKK3.

DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DKK3
Polyclonal
Unconjugated
DKK3
AW061014; C87148; Dickkopf; dickkopf 3; dickkopf 3 homolog; Dickkopf gene 3; dickkopf homolog 3; dickkopf homolog 3 (Xenopus laevis); dickkopf WNT signaling pathway inhibitor 3; Dickkopf3; dickkopf-3; Dickkopf-related protein 3; DKK 3; Dkk3; dkk-3; hDkk 3; hDkk-3; hhDkk3; mDkk-3; p29; regulated in glioma; Reic; RIG; RIG-like 5-6; RIG-like 7-1; UNQ258/PRO295; Xenopus
Rabbit
Antigen affinity chromatography, Protein A
RUO
27122
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Paraffin)
1 mg/mL
PBS with no preservative
Q9UBP4
DKK3
Recombinant Human DKK3 Protein.
100 μL
Primary
Human
Antibody
IgG
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.