Learn More
Ditolylguanidine, MedChemExpress
Product Code. 16471399
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16471399 Supplier Medchem Express Supplier No. HY14218
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ditolylguanidine (1,3-Di-o-tolylguanidine) is an agonist of sigma receptor (σ1/σ2 receptor).
Ditolylguanidine (1,3-Di-o-tolylguanidine) is an agonist of sigma receptor (σ1/σ2 receptor).
Specifications
Chemical Name or Material Ditolylguanidine
Physical Form Solid
CAS 97-39-2
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C15H17N3
For Use With (Application) Neuroscience-Neuromodulation
Synonym 1,3-Di-o-tolylguanidine DTG
Solubility Information DMSO : 150 mg/mL (626.78 mM; Need ultrasonic)
SMILES CC1=C(NC(NC2=CC=CC=C2C)=N)C=CC=C1
Molecular Weight (g/mol) 239.32
Formula Weight 239.32
Percent Purity 99.03%
Grade Research