Learn More
Disufenton sodium, MedChemExpress
Product Code. 16491747
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16491747
Brand: Medchem Express HY13244
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Disufenton sodium (NXY-059) is the disulfonyl derivative of the neuroprotective spin trap phenylbutynitrone(PBN), both NXY-059, its parent PBN and their hydrolysis/oxidation product MNT are very powerful scavengers of free radicals.
Disufenton sodium (NXY-059) is the disulfonyl derivative of the neuroprotective spin trap phenylbutynitrone(PBN), both NXY-059, its parent PBN and their hydrolysis/oxidation product MNT are very powerful scavengers of free radicals.
Specifications
Chemical Name or Material Disufenton sodium
CAS 168021-79-2
Color White
Purity Grade Notes Research
Molecular Formula C11H13NNa2O7S2
Quantity 50 mg
Synonym NXY-059
Solubility Information DMSO : 125 mg/mL (327.80 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (131.12 mM)
SMILES O=S(C1=C(C=CC(S(=O)(O[Na])=O)=C1)/C=[N+]([O-])/C(C)(C)C)(O[Na])=O
Molecular Weight (g/mol) 381.33
Formula Weight 381.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration