Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz-Thiele, Bent
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05298001
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10623242
  • 265.00 € / Each
Estimated Shipment: 09-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Receivers with bent adapter, graduated
  • With NS socket and NS cone
  • vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifications
Specifications
Bent adapter, graduated, with NS socket and NS cone, vacuum connection: thread GL18
Duran Glass
25 mL
Socket and Cone NS 14/23