Learn More
Greiner Bio-One Disposable Inoculating Loops
Product Code. 10240921
Click to view available options
Volume:
1 μL
10 μL
Color:
Blue
White
Packaging Type:
10/bag
Unit Size:
Pack of 3000
Product Code. 10240921
Brand: Greiner Bio-One 731175
Only null left
Add to basket
Add to basket
For use in picking single colonies. Greiner Bio-One Disposable Inoculating Loops are flexible for collection and inoculation.
  • Sterilised by irradiation
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 10 μL
Length (Metric) 200 mm
Material Polystyrene
Sterility Sterile
Color Blue
Quantity 3000
Packaging Type 10/bag