Learn More
Greiner Bio-One Disposable Inoculating Loops
Click to view available options
Volume:
1 μL
10 μL
Color:
Blue
White
Packaging Type:
10/bag
Unit Size:
Pack of 3000
Description
- Sterilised by irradiation
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Length (Metric) | 200 mm |
| Material | Polystyrene |
| Sterility | Sterile |
| Color | White |
| Quantity | 3000 |
| Packaging Type | 10/bag |