Learn More
Sartorius™ Disposable Chute Bags
Shop All Sartorius Products
Click to view available options
:
Disposable Chute Bags; 100/Pk
Description
Safety weighing cabinets ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing of toxic and high-potency samples.
- Designed for use with SWC Safety Weighing Cabinets
- Disposal chute attaches securely to the side of the cabinet
- Disposable chute bags enable safe disposal of substances without exposure to the lab
- Contaminated materials can be disposed of safely in a double-bag system
- Packaged: 100/Pack
Specifications
Specifications
| For Use With (Equipment) | YWCG02 Disposal Chute for Waste, SWC Safety Weighing Cabinet |
| Color | Transparent |
| For Use With (Application) | Waste disposal |
| Quantity | 100/Pk. |
| Product Line | SWC |