Learn More
Brady™ Disposable Chemical Spill Kit
Product Code. 15995171 Shop All Brady Products
Change view
Click to view available options
Color: Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15995171 Color: Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15995171 Supplier Brady™ Supplier No. 813868
  • ONLINE EXCLUSIVE PRICE
    121.50 €
    100.99 € / Pack of 4
    Save 20.51 €
    16% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Portable spill kit that comes in a green, disposable dispenser box.
  • Portable
  • Disposable
  • Designed for small spills of chemicals and hazardous liquids
  • Includes an autoclavable disposal bag that is heat resistant up to 145°C
  • For safe disposal of laboratory waste
Specifications
Color Green
Type Chemical Sorbent
Includes 2 pads (40 x 50 cm), 1 pair of disposable chemical protective gloves, 1 autoclavable disposal bag, 1 disposable brush, and multilingual instruction guide
Alternate Part Number SKH-MINI