Learn More
Kinematica Dispersing Aggregate Dia. 60mm PT-DA 60/2EC-F250
Brand:  Kinematica 11030338
Dispersing Aggregate Dia. 60mm PT-DA 60/2EC-F250
Product Code. 15586929
  • ONLINE EXCLUSIVE PRICE
    4121.25 €
    3825.00 € / Each
    Save 296.25 €
    7% Off
Only null left
Estimated Shipment: 27-01-2026