missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DISP1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA548032
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
DISP1 Polyclonal antibody specifically detects DISP1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
DISP1 | |
Polyclonal | |
Unconjugated | |
DISP1 | |
1190008H24Rik; chameleon; con; Disp; Disp1; Dispa; dispatched 1; dispatched A; dispatched homolog 1; dispatched homolog 1 (Drosophila); dispatched RND transporter family member 1; DKFZP434I0428; FLJ43740; helix; hlx; hypothetical protein LOC619201; Icb; Icbins; Mdispa; MGC104180; MGC13130; MGC16796; multipass transmembrane protein; Disp1; protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein; zgc:111866 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
84976 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96F81 | |
DISP1 | |
E. coli-derived recombinant human DISP1 Glu212-Lys492, Ser746-Thr984 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |