missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dishevelled-2 Antibody (OTI2D11), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70611PE
This item is not returnable.
View return policy
Description
Dishevelled-2 Monoclonal antibody specifically detects Dishevelled-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Dishevelled-2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI2D11 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| dishevelled 2 (homologous to Drosophila dsh), dishevelled, dsh homolog 2 (Drosophila), Dishevelled-2, DSH homolog 2, segment polarity protein dishevelled homolog DVL-2 | |
| Human recombinant protein fragment corresponding to amino acids 1-320 of human DVL2(NP_004413) produced in E.coli. | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 1856 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction