Learn More
B Braun™ Discofix™ 3-Way Stopcock
Lock swivels both axially and radially for secure and speedy connection. B Braun™ Discofix™ 3-Way Stopcock is suitable for all current pressure infusion systems.
Brand:  B Braun™ 4095111
Additional Details : Weight : 0.00540kg
Product Code. 10199361
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Stopcock system for infusion therapy and monitoring
  • Suitable for all current pressure infusion systems
  • Continuous straight-flow channels for accurate pressure monitoring
  • Swivel lock moves both axially and radially for secure and speedy connection