Learn More
Hirschmann™ Discharge Tube Unit Titration
Product Code. 11322513
Change view
Click to view available options
Material:
FEP
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
11322513 FEP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11322513 Supplier Hirschmann™ Supplier No. 9313041
Only null left
Add to basket
Add to basket
Dispensing nozzle with FEP-tubing and fine tapered tip for use with solarus™ and opus™ titration (10 mL instruments).
  • Guided tubing
  • Screw-coupled
  • With closure cap
Specifications
Description Discharge unit titration with fine tip
Includes 1
Material FEP