Learn More
Hirschmann™ Discharge Tube Unit Dispensing
Product Code. 10438222
Change view
Click to view available options
:
Product Type: Discharge Tube Unit Complete
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10438222 Product Type: Discharge Tube Unit Complete
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10438222 Supplier Hirschmann™ Supplier No. 9313025
Only null left
Add to basket
Add to basket
Dispensing nozzle with FEP-tubing for use with ceramus™ and opus™ dispenser.
  • Guided tubing
  • Screw-coupled
  • With closure cap
Specifications
Quantity 1 Each
Type Discharge Tube Unit
For Use With (Equipment) ceramus™
Material FEP
Description With completely guided tube