Learn More
DIRC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567249
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: IPISDLILKRR LIHGGQMLNG LAGPTVMNAA PFLSTTWFSA DERATATAIA S Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Electrogenic metabolite transporter.Specifications
DIRC2 | |
Polyclonal | |
Unconjugated | |
Dirc2 | |
Dirc2; Disrupted in renal cancer protein 2; disrupted in renal carcinoma 2; disrupted in renal carcinoma 2 (human); disrupted in renal carcinoma 2 homolog; disrupted in renal carcinoma protein 2; Disrupted in renal carcinoma protein 2 homolog; RCC4; renal cell carcinoma 4; SLC49A4; Solute carrier family 49 member 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96SL1 | |
Dirc2 | |
Recombinant Human DIRC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only