missing translation for 'onlineSavingsMsg'
Learn More

Diquafosol tetrasodium, MedChemExpress

Product Code. p-200096471
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488548

Brand: Medchem Express HYB0606

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.

Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.

Specifications

Chemical Name or Material Diquafosol tetrasodium
CAS 211427-08-6
Color White
Purity Grade Notes Research
Molecular Formula C18H22N4Na4O23P4
Quantity 10 mg
Synonym INS365
Solubility Information H2O : 100 mg/mL (113.87 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.14 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](N(C=CC2=O)C(N2)=O)O[C@H](COP(OP(OP(OP(OC[C@H]3O[C@@H](N(C=CC4=O)C(N4)=O)[C@H](O)[C@@H]3O)(O[Na])=O)(O[Na])=O)(O[Na])=O)(O[Na])=O)[C@H]1O
Molecular Weight (g/mol) 878.23
Formula Weight 878.23
Percent Purity 98.49%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.