missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipyrone Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA175123
Additional Details : Weight : 0.01000kg
Description
PA1-75123 specifically detects Dipyrone. PA1-75123 has been successfully used in ELISA applications. The PA1-75123 immunogen is Dipyrone(4-methylamino)-BTG.
Dipyrone is a drug that has analgesic, anti-inflammatory, and antipyretic properties. It is the sodium sulfonate of AMINOPYRINE. Metamizole, formerly marketed as Dimethone tablets and injection, Protemp oral liquid, and other drug products, was associated with potentially fatal agranulocytosis. Approvals of the NDA's for dipyrone drug products were withdrawn on June 27, 1977. [National Center for Biotechnology Information. PubChem Compound Database; CID=522325].Specifications
Dipyrone | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
Analgin; C13H16N3NaO4S; Metamizole sodium; Methampyrone; Sulpyrine | |
Dipyrone(4-methylamino)-BTG | |
1 mg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |