missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride

Product Code. p-7042984
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426609 50 mg 50mg
15416609 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426609 Supplier Thermo Scientific Alfa Aesar Supplier No. J64838.MF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4673-26-1 | C12H8ClI | 314.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
MDL Number MFCD00214165
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Melting Point >310°C
Color White
Quantity 50 mg
Formula Weight 314.55
Physical Form Powder
Chemical Name or Material Diphenyleneiodonium chloride
compliance-icons
Product Identifier
  • Diphenyleneiodonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Diphenyleneiodoniumchloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.