Learn More
Diphenylcarbamyl chloride, 98%
Product Code. 11416336
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416336 100 g 100g
11406336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11416336 Supplier Thermo Scientific Alfa Aesar Supplier No. B23116.22
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
CAS: 83-01-2 | C13H10ClNO | 231.679 g/mol
Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83-01-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.679
MDL Number MFCD00000633
InChI Key XNBKKRFABABBPM-UHFFFAOYSA-N
Synonym diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
PubChem CID 65741
IUPAC Name N,N-diphenylcarbamoyl chloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl
Specifications
Melting Point 82°C to 86°C
UN Number UN3261
Beilstein 515312
Solubility Information Reacts with water.
Quantity 100 g
Odor Irritating
Formula Weight 231.68
Sensitivity Moisture Sensitive
Percent Purity 98%
Chemical Name or Material Diphenylcarbamyl chloride