missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcarbamyl chloride, 98%

Product Code. p-7026851
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406336 25 g 25g
11416336 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406336 Supplier Thermo Scientific Alfa Aesar Supplier No. B23116.14
Add to basket
This item is not returnable. View return policy

CAS: 83-01-2 | C13H10ClNO | 231.679 g/mol

Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-01-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.679
MDL Number MFCD00000633
InChI Key XNBKKRFABABBPM-UHFFFAOYSA-N
Synonym diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
PubChem CID 65741
IUPAC Name N,N-diphenylcarbamoyl chloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 82°C to 86°C
UN Number UN3261
Beilstein 515312
Solubility Information Reacts with water.
Quantity 25 g
Odor Irritating
Formula Weight 231.68
Sensitivity Moisture Sensitive
Percent Purity 98%
Chemical Name or Material Diphenylcarbamyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.