missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphoramidate, 97%

Product Code. p-7044567
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347156 5 g 5g
11357156 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347156 Supplier Thermo Scientific Alfa Aesar Supplier No. L07155.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2015-56-7 | C12H12NO3P | 249.206 g/mol

Diphenyl phosphoramidate is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphoramidate is used as primary and secondary intermediates.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2015-56-7
Molecular Formula C12H12NO3P
Molecular Weight (g/mol) 249.206
MDL Number MFCD00014078
InChI Key QWMUDOFWQWBHFI-UHFFFAOYSA-N
Synonym diphenyl phosphoramidate, diphenyl amidophosphate, phosphoramidic acid, diphenyl ester, diphenylphosphoramidate, amino phenoxy phosphoryl oxy benzene, diphenyl phosphoroamidate, aminodiphenoxyphosphino-1-one, amidophosphoric acid, diphenyl ester, acmc-1cki3
PubChem CID 74836
IUPAC Name [amino(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(N)OC2=CC=CC=C2

Specifications

Melting Point 147°C to 151°C
Quantity 5 g
Beilstein 2054166
Solubility Information Insoluble in water.
Formula Weight 249.21
Percent Purity 97%
Chemical Name or Material Diphenyl phosphoramidate

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.