missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl(1,5-cyclooctadiene)platinum(II), Thermo Scientific Chemicals

Product Code. 11420660
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420660 5 g 5g
11410660 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420660 Supplier Thermo Scientific Alfa Aesar Supplier No. 044039.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12277-88-2
Molecular Formula C20H22Pt
Molecular Weight (g/mol) 457.48
MDL Number MFCD03788255
InChI Key MQCNDJYHICNUPD-PHFPKPIQSA-N
Synonym 1,5-cyclooctadiene, z,z-; diphenylplatinum
PubChem CID 92045412
IUPAC Name benzene;(5Z)-cycloocta-1,5-diene;platinum(2+)
SMILES C1C\C=C/CC\C=C/1.[Pt++](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 457.47
Physical Form Powder
Chemical Name or Material Diphenyl (1,5-cyclooctadiene)platinum (II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.